missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex™ 12% Tris-Glycine Protein Gel, 20-well, w/adapters
Click to view available options
Wells:
20-well
Unit Size:
Pack of 10
Description
Includes
One box of 10 gels, adapters
- Provide reproducible separation of a wide range of proteins into well-resolved bands
- Individually packaged gels for optimal experiment design
- Compatible with most protein standards
- Usable for native and denaturing protein assays
- Do not contain SDS and can be used to run proteins in native or in denatured form
Sample Loading Volume: 25μL; 1D Well
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Gel Percentage | 12% |
| Gel Type | Tris-Glycine |
| Product Type | Homogeneous Page |
| Separation of | Protein |
| Separation Range | 20 to 120 kDa |
| Wells | 20-well |
| Quantity | 1 Box |
| Well Design | 1D Well |
| Length (Metric) | 8 cm |
| Width (Metric) | 13 cm |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction