missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABTS, solution

Product Code. 15949122
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15949122 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15949122 Supplier Invitrogen™ Supplier No. 002024
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suitable for use in immunoassays

May be used as a substrate for horseradish peroxidase (HRP) in ELISAs.

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2-8°C
Detection Method Colorimetric
Product Type ABTS Solution
Substrate ABTS
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase) Substrate
Quantity 500 mL
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.