missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AcTEV™ Protease

Product Code. 10216572
Change view
Click to view available options
Quantity:
10,000 U
1000 U
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10216572 1000 U Each
10443432 10,000 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10216572 Supplier Invitrogen™ Supplier No. 12575015
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

AcTEV™ Protease is supplied with a vial of 20X TEV buffer (1M Tris-HCl (pH 8.0), 10mM EDTA) and a vial of 100mM DTT.

Specifically recognizes a seven amino acid sequence (Glu-Asn-Leu-Tyr-Phe-Gln-Gly, cleaving between Gln and Gly), ideal for removing affinity tags from fusion proteins

An improved version of Tobacco Etch Virus (TEV) AcTEV™ Protease is highly site-specific, highly active and significantly more stable than native TEV protease, resulting in enhanced long-term activity.

  • Highly specific cleavage activity
  • Enhanced enzyme stability for prolonged protease activity
  • Activity over a broad temperature (4°C to 30°C) and pH (6.0 to 8.5) range
  • Six-histidine sequence to facilitate its removal from the digested protein sample
  • Greater than 85% single-band purity with no nonspecific protease contamination

  • Application
    • Incubation with AcTEV™ Protease releases the protein of interest from the fusion tag
    • This is an effective way to remove solubility, secretion, detection, and purification tags from recombinant protein
    Enzyme specifications
    • Purified from E. coli expressing the AcTEV™ Protease gene
  • Unit definition
    • One unit of AcTEV™ Protease cleaves 85% of a 3μg control substrate in 1 hr at 30°C
  • Unit reaction conditions
    • 50mM Tris-HCl (pH 8.0), 0.5mM EDTA, 1mM DTT, 3μg control substrate, and 1 unit enzyme in 30μL for 1 hr at 30°C AcTEV™ Protease is functionally tested for the absence of any nonspecific protease activity

    GST-Tagged Protein Purification, His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1,000 Units
Content And Storage AcTEV™ Protease is supplied with a vial of 20X TEV buffer [1 M Tris-HCl (pH 8.0), 10 mM EDTA] and a vial of 100 mM DTT.

Store at -20° C. Guaranteed stable for 1 year when properly stored.
Shipping Condition Dry Ice
Enzyme TEV Protease
Compatible Buffer TEV Buffer
Optimal Reaction Temperature +4°C to +30°C
Quantity 1000 U
Product Type Protease
Product Line AcTEV
Why does AcTEV Protease require DTT?

A final concentration of 1 mM DTT is required for the AcTEV protease reaction. The DTT serves as a stabilizer of secondary structure, i.e., it ensures that the enzyme does not undergo oxidation. If you are unable to use DTT due to a column purification after digestion, you can leave it out and still see a successful AcTEV digestion, however we recommend using it.

No rigorous quantitative data for activity in the absence of DTT and EDTA has been collected in-house. Incubations using reaction buffer +/-DTT and +/- EDTA; have been performed, but the products were analyzed only at t=0 and t=2 hr at 30 degrees C. No difference in the amount of product generated +/-DTT or +/-EDTA was noticed, but it is possible that the detailed kinetics in a time course reaction may be a little different.

What is the cleavage recognition site for AcTEV Protease?

The recognition site for AcTEV Protease is ENLYFQS/G

What products do you offer for enzymatic cleavage of fusion tags from recombinant proteins?

We offer the following products:

-AcTEV Protease (Cat. Nos. 12575015, 12575023)
-EKMax Enterokinase (Cat. Nos. E18001, E18002)
-SUMO Protease (Cat. No. 12588018)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.