missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bolt™ Transfer Buffer (20X)

Product Code. 17891041
Change view
Click to view available options
Quantity:
125mL
1L
Unit Size:
1 L
125mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17891041 1L 1 L
15135204 125mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17891041 Supplier Invitrogen™ Supplier No. BT00061
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Requires

A Bolt™ Mini Gel Tank is required for use of all Bolt gels.

Optimized for the transfer of proteins from Bolt Bis-Tris Plus gels to membranes for western blotting

  • When combined with 10% methanol, Bolt Transfer Buffer can be used with Bolt Mini Blot Module for wet transfer
  • Can also be used with Bolt Antioxidant to enhance transfer of reduced proteins to membranes

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4 to 25°C.
Buffer Transfer Buffers
Concentration 20 X
Volume (Metric) 1 L
Form Liquid
Gel Compatibility Bolt™ Bis-Tris Plus Gels
Quantity 1L
Product Line Bolt
Product Type Western Blot Transfer Buffer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.