missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EKMax™ Enterokinase

Product Code. 10453242
Change view
Click to view available options
Quantity:
1000 Units
250 Units
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10453242 1000 Units Each
10184792 250 Units Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10453242 Proveedor Invitrogen™ N.º de proveedor E18002

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Includes

EKMax Enterokinase is provided with 10X buffer.

Recombinant preparation of the catalytic subunit of bovine enterokinase

  • Recognizes the sequence Asp-Asp-Asp-Asp-Lys and cleaves peptide bond after lysine residue
  • Enzyme can be used to cleave any fusion protein that carries this peptide sequence
  • Well-suited to removal of fusion tags from recombinant proteins
  • Unit Definition: One unit of EKMaxis amount of enzyme required to digest 20μg of thioredoxin-CAT fusion protein to 90% completion in 16 hours at 37°C; One EKMaxunit is equivalent to ∽190 trypsinogen activation units

GST-Tagged Protein Purification, His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Especificaciones

Product Type Enterokinase
Content And Storage EKMax™ Enterokinase is provided with 10X buffer. Store at -20°C. Guaranteed stable for 6 months when stored properly.
Species Cattle/Bovine
Quantity 1000 Units
Product Line EKMax

For Research Use Only. Not for use in diagnostic procedures.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.