Learn More
Invitrogen™ Human FGF2 ELISA Kit
Description
Includes
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
The Human FGF-b ELISA Kit quantifies human Fibroblast Growth Factor basic (FGF-b) in serum, plasma, buffered solution, or cell culture medium. The assay will recognize both the natural 18 kDa isoform and recombinant FGF basic-146 and FGF basic-157. Principle of the method The Human FGF-b solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Accession Number | P09038 |
| Assay Range | 15.6 to 1000 pg/mL |
| Assay Sensitivity | 7 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Application) | Colorimetric Detection |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 2247 |
| Show More |
Safety and Handling
- Human FGF2 ELISA Kit
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : SODIUM AZIDE
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.