missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Hi-Density TBE Sample Buffer (5X)

Product Code. 12070146
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12070146 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 12070146 Supplier Invitrogen™ Supplier No. LC6678

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Compatible with polyacrylamide gels

Novex TBE Gels provide the highest resolution possible for analyzing restriction digests and PCR products. DNA fragments ranging from 10 to 3,000 base pairs are clearly resolved into sharp, tight bands. Novex DNA Retardation gels are prepared with 0.5X TBE as the gel buffer. This is sufficient for good electrophoretic separation, yet low enough to promote DNA-protein interactions

  • Novex TBE and DNA Retardation Gels are designed to run on XCell SureLock™ Mini-Cell
  • Formulation: Novex TBE and DNA Retardation Gels are manufactured with high-purity tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS
  • Recommended Buffers: For optimum performance, Novex TBE Running Buffer and Novex Hi-Density TBE Sample Buffer are strongly recommended for use with these gels
  • Novex Hi-Density TBE Sample Buffer contains the density agent Ficoll™, which yields sharper, straighter bands than conventional density agents, and tracking dyes Bromophenol Blue and Xylene Cyanol

Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Buffer Sample Loading Buffer
Concentration 5 X
Content And Storage Store at 2°C to 8°C
Gel Compatibility Polyacrylamide Gels
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Label or Dye Bromophenol Blue, Xylene Cyanol
Product Line Novex
Product Type TBE Buffer
Quantity 10 mL
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.