Learn More
Invitrogen™ IFN gamma Mouse ELISA Kit
Quantifies natural and recombinant mouse IFN γ in serum, plasma, buffered solution, or cell culture medium
668.00 € - 2145.00 €
Specifications
Accession Number | P01580 |
---|---|
Assay Range | 47-300 pg/mL |
Assay Sensitivity | <2 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10078932
|
Invitrogen™
KMC4021 |
96 Tests |
668.00 €
Each |
In Stock
Log in to see stock. |
|||||
10486483
|
Invitrogen™
KMC4022 |
2 x 96 Tests |
998.00 €
Each |
In Stock
Log in to see stock. |
|||||
10543205
|
Invitrogen™
KMC4021C |
5 x 96 Tests |
2145.00 €
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
Description
The Mouse Interferon Gamma (Ms IFNγ) ELISA quantitates Ms IFNγ in mouse serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IFNγ. Principle of the method The Mouse IFNγ solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Interferon-gamma (IFN γ) is a lymphokine produced by activated T and NK cells.
Cell Analysis, Cell Signaling, Cell Viability, Proliferation & Function, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Room Temperature
Specifications
P01580 | |
<2 pg/mL | |
ELISA Kit | |
Mouse | |
Colorimetric Microplate Reader | |
IFN-g,Ifg | |
6.5% | |
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |
47-300 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Colorimetric Detection | |
15978 | |
4 hr. | |
3.2% | |
HRP | |
Plasma, 25 μL; Serum, 25 μL; Supernatant, 25 μL | |
Mouse | |
4 hr. |
For Research Use Only. Not for use in diagnostic procedures.