missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lumio™ Green Detection Kit

Product Code. 10318402
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10318402 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10318402 Supplier Invitrogen™ Supplier No. LC6090

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

The Lumio™ Green Detection Kit contains one vial of Lumio™ Green Detection Reagent, five vials of Lumio™ Gel Sample Buffer, and one vial of Lumio™ In-Gel Detection Enhancer.

Enables immediate visualization of Lumio™ fusion protein bands directly in a polyacrylamide gel

The Lumio™ Green Detection Kit is a sensitive and highly specific kit for labeling Lumio™ fusion proteins prior to electrophoresis.

  • Label proteins fused to a Lumio sequence
  • Visualize results immediately after electrophoresis, eliminate gel staining and western blotting
  • Detect nanogram levels of protein with UV transilluminator, equipped with standard camera (Figure 1), or laser-based scanner

Enzyme and Protein Activity Assays, Lumio Detection, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis, Reporter Assays

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The Lumio™ Green Detection Kit contains one vial of Lumio™ Green Detection Reagent, five vials of Lumio™ Gel Sample Buffer, and one vial of Lumio™ In-Gel Detection Enhancer. Store at -20°C. All components are guaranteed stable for 6 months when properly stored.
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye Lumio Green
Quantity 1 kit
Shipping Condition Dry Ice
Target Molecule Proteins (Lumio™-Tagged) , Proteins (TC-Tagged)
Product Line Lumio
Product Type Pre-Gel Fusion Protein Labeling Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.