missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Protein Molecular Weight Standards (broad range)

Product Code. 10063252
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10063252 400 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10063252 Supplier Invitrogen™ Supplier No. P6649

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Include 11 proteins from 6500 to 205,000 daltons

The broad range protein molecular standards for use as molecular weight standards in SDS-polyacrylamide gel electrophoresis.

  • Novex Tricine Gels, Novex Tris-Glycine Gels, NuPAGE Bis-Tris Gels, NuPAGE Tris-Acetate Gels, SDS-PAGE Gels

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Number of Markers 11
Ready to Load No
Sample Loading Volume 2 μL
Size Range 6.5 to 205 kDa
Gel Compatibility Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol) 205, 116, 97, 80, 66, 55, 45, 30, 21, 14, 6.5 kDa
Quantity 400 μL
Shipping Condition Wet Ice
Product Type Protein Molecular Weight Standard
Stain Type Unstained
System Type SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.