missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SimplyBlue™ SafeStain

Product Code. 10533053
Change view
Click to view available options
Quantity:
1 L
3.5 L
Unit Size:
1 L
3.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10533053 3.5 L 3.5 L
10432072 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10533053 Supplier Invitrogen™ Supplier No. LC6065
Spotlight Offers
Use promo code 28037
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

SimplyBlue™ SafeStain is supplied as a 1X ready-to-use staining reagent.

SimplyBlue SafeStain is a ready-to-use, fast, sensitive, and safe Coomassie G-250 stain for visualizing protein bands on polyacrylamide gels and on dry PVDF membranes.

SimplyBlue SafeStain is a ready-to-use, fast, sensitive, and safe Coomassie G-250 stain for visualizing protein bands on polyacrylamide gels and on dry PVDF membranes. SimplyBlue SafeStain is completely non-hazardous and does not require methanol or acetic acid fixatives or destains. Risk of hazardous exposure and unpleasant odors are eliminated, creating a healthier lab environment.

Coomassie G-250 stain for visualizing protein bands on polyacrylamide gels and on dry PVDF membranes. SimplyBlue SafeStain is completely non-hazardous and does not require methanol or acetic acid fixatives or destains. Risk of hazardous exposure and unpleasant odors are eliminated, creating a healthier lab environment.

Features include:
• An easy-to-perform protocol that can be completed in less than three hours. For added speed, a simple microwave procedure can be completed in 12 minutes
• Destaining not required but may be performed with deionized water to achieve maximum sensitivity, especially when performing downstream analyses such as mass spectrometry, or when a crystal-clear background is required
• Sensitive—detect down to 7 ng of protein
• Fast—detect 1 μg of protein in as little as five minutes after staining
• Convenient—save valuable time by skipping reagent preparation with this ready-to-use formulation
• Non-hazardous—save money and hassle associated with hazardous shipping and disposal of hazardous waste

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage SimplyBlue™ SafeStain is supplied as a 1X ready-to-use staining reagent. Store at room temperature. Guaranteed stable for 6 months when properly stored. There are no HazMat charges associated with this product.
Detection Location In-Gel Detection
Color Blue
Detection Method Colorimetric
Label or Dye Coomassie
Quantity 3.5 L
Target Molecule Protein
Product Line SimplyBlue
Product Type Safe Protein Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.