missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SUMO Protease

Product Code. 10338632
Change view
Click to view available options
Quantity:
250 units
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10338632 250 units Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10338632 Supplier Invitrogen™ Supplier No. 12588018
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

SUMO Protease is provided with 10X high salt and 10X no salt dilution buffers.

A recombinant fragment of ULP1 (Ubl-specific protease 1) from Saccharomyces cerevisiae

SUMO Protease, also known as Ulp, is highly specific for the SUMO protein fusion, recognizing the tertiary structure of SUMO rather than an amino acid sequence.

  • Highly active cleavage
  • No non-specific proteolysis (highly specific cleavage)
  • Activity from 2°C to 37°
  • A six-histidine sequence to facilitate its removal from the digested protein sample

Application:Removal of fusion tags from recombinant proteins
Unit Definition: One unit of SUMO Protease is defined as the amount of enzyme needed to cleave 85% of 2μg of substrate protein at 30°C in one hour
Quality Control: SUMO Protease has greater than 85% single-band purity with no non-specific protease contamination; functionally tested for the absence of any non-specific protease activity

GST-Tagged Protein Purification, His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Host Species S. cerevisiae
Content And Storage SUMO Protease is provided with 10X high salt and 10X no salt dilution buffers. Store at -80°C. Guaranteed stable for 6 months when properly stored.
Shipping Condition Dry Ice
Quantity 250 units
compliance-icons
Product Identifier
  • SUMO Protease
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Nonidet P-40

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.