missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ TBE Running Buffer (5X)

Product Code. 10247152
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10247152 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10247152 Supplier Invitrogen™ Supplier No. LC6675

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Compatible with agarose and polyacrylamide gels

Novex™ TBE Gels provide highest resolution possible for analyzing restriction digests and PCR products. DNA fragments ranging from 10 to 3,000 base pairs are clearly resolved into sharp, tight bands.

  • Prepared with 0.5X TBE as gel buffer
  • Sufficient for good electrophoretic separation, yet low enough to promote DNA-protein interactions
  • TBE and DNA Retardation Gels are designed to run on the XCell SureLock™ Mini-Cell

Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Buffer Running Buffer
Concentration 5 X
Content And Storage Store at room temperature
Gel Compatibility Polyacrylamide Gels, Agarose Gels
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Product Line Novex
Product Type TBE Running Buffer
Quantity 1 L
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.