missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ TBE-Urea Gels, 15%

Product Code. p-7250387
Change view
Click to view available options
Wells:
10-well
12-well
15-well
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells unitSize
12040126 10-well Pack of 10
12050126 12-well Pack of 10
12060126 15-well Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12040126 Supplier Invitrogen™ Supplier No. EC6885BOX

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Catalog numbers for gels refer to a single box. One box contains 10 gels. Novex TBE-Urea Gels have a shelf life of between 4 and 8 weeks depending upon gel type.

Novex™ TBE-Urea Gels are denaturing polyacrylamide gels that resolve single-stranded DNA oligos or RNA into sharp, distinct bands.

Novex™ TBE-Urea Gels are denaturing polyacrylamide gels that resolve single-stranded DNA oligos or RNA into sharp, distinct bands. These 15% gels are optimized for the analysis and purification of products ranging from 10–40 bases, making them an ideal choice for synthetic oligo analysis and purification, RNase Protection Assays (RPA), in vitro transcription studies, and northern blot analysis.

Novex TBE-Urea Gels can be stained by silver staining, ethidium bromide, and SYBR™ Green staining techniques after electrophoresis. The compact gels are available in 6%, 10%, or 15% formats and are designed to run on the XCell SureLock™ Mini-Cell.

Advantages of TBE-Urea Gels:
• Contains 7M urea for maximum denaturation
• Excellent resolution for fast size and purity confirmations of DNA or RNA oligos
• High resolution of short single-strand oligonucleotides
• Requires ∼10% sample concentration and volume of large or agarose gels
• Accurate and reproducible results

Formulation
Novex TBE-Urea gels are made with high-purity reagents, including Tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, APS, and 7M urea. Strict quality control ensures that gels and buffers are DNase- and RNase-free.

Recommended Buffers
For optimum performance, Novex™ TBE-Urea sample buffers and Novex™ TBE Running Buffer are strongly recommended for use with these gels. For analytical applications, Novex TBE-Urea Sample Buffer is recommended contains urea, the density agent Ficoll™, which yields sharper, straighter bands than conventional density agents, and the tracking dyes Bromophenol Blue and Xylene Cyanol.

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Catalog number refers to a single box of 10 gels
• Novex™ TBE-Urea Gels have a shelf life of 4 to 8 weeks depending upon gel type

Store at 4°C.
Description Novex TBE-Urea Gels, 15%, 10-well
Gel Percentage 15%
Gel Type TBE-Urea
Separation Range 10 to 40 bp
Wells 10-well
Quantity 10 Gels/Box
Sample Type Single-stranded DNA Oligos/RNA
Shipping Condition Wet Ice
Product Type TBE Gel
For Use With (Equipment) XCell SureLock Mini-Cell
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.