Learn More
Invitrogen™ Novex™ TBE-Urea Sample Buffer (2X)
Compatible with agarose and polyacrylamide gels
Brand: Invitrogen™ LC6876
Code : 50
Additional Details : Weight : 0.00410kg
Description
For optimum performance, Novex™ TBE-Urea Sample Buffer is recommended for use with Novex™ TBE-urea gels.
- Contains urea and density agent Ficoll™
- Yields sharper, straighter bands than conventional density agents, plus tracking dyes bromophenol blue and xylene cyanol
Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting
Order Info
Shipping Condition: Wet Ice
Specifications
Sample Loading Buffer | |
Store at 2°C to 8°C | |
Nucleic Acid Gel Electrophoresis, Blotting | |
Novex™ | |
10 mL |
2 X | |
Polyacrylamide Gels, Agarose Gels | |
Bromophenol Blue, Xylene Cyanol | |
TBE Sample Buffer | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.