missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELISA TMB Stabilized Chromogen

Product Code. 10523405
Change view
Click to view available options
Quantity:
1 L
25 mL
Unit Size:
1 L
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10523405 25 mL 25mL
10354603 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10523405 Supplier Invitrogen™ Supplier No. SB01
 Restricted Item
In Stock
Add to basket
Edge
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

For use with all sandwich ELISAs which utilize Horseradish Peroxidase (HRP) as detecting enzyme

  • Formulation: Liquid
  • Available in two sizes
  • Detection method: Colormetric

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifica

Quantity 25 mL
Content And Storage Solution. Store at 2-8°C.
Detection Method Colorimetric
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.