Learn More
Invitrogen™ WesternBreeze™ Chemiluminescent Kit, anti-mouse
Description
Detection is accomplished with a ready-to-use CDP-Star™ chemiluminescent substrate for alkaline phosphatase. Protein bands can be captured either by X-ray film or CDP-Star™ compatible imaging system.
WesternBreeze™ Chemiluminescent Kit offers:
- High specificity, clean background
- Ultra-sensitivity-femtogram levels detectable
- Long-lasting signals-up to 5 days
- Results in less than 3 hours
- Permanent photographic image
Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
Specifications
| Product Type | Western Blot Kit |
| Content And Storage | The WesternBreeze™ Chemiluminescent Kits include blocking solutions, primary antibody diluent, ready-to-use secondary antibody solution (anti-mouse, anti-rabbit or anti-goat), ready-to-use chemiluminescent substrate, wash solutions, incubation trays, pre-cut filter papers, polyester sheet for even substrate development on the membrane. Each kit contains complete reagents for 20 blots. Store the kits at +4°C. Guaranteed stable for 6 months when properly stored. |
| Reactivity | Mouse |
| Shipping Condition | Wet Ice |
| Substrate Type | AP (Alkaline Phosphatase) Substrate |
| Detection Method | Chemiluminescence |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Product Line | WesternBreeze |
| Cross Reactivity | Mouse |
| Quantity | 1 kit |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.