missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sponge Pad for Blotting

Product Code. 11554756
Change view
Click to view available options
Quantity:
8 pads
Unit Size:
Pack of 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11554756 8 pads Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11554756 Supplier Invitrogen™ Supplier No. EI9052
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the XCell SureLock Mini Blot Module

Applications:

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Application) 1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
For Use With (Equipment) XCell SureLock™ Mini
Gel Size Mini
Product Line XCell II
Type Sponge Pad
Quantity 8 pads
Shipping Condition Room Temperature
What are the dimensions of the sponge pads you offer for blotting?

The dimensions of the sponge pads are 8 cm x 9 cm.

Do you offer replacement parts for the XCell II Blot Module?

Except for the sponge pads (Cat. No. EI9052), none of the other parts are available separately.

Should I clean the sponge pads after each use and how should I do it?

We recommend cleaning the sponge pads after each use by rinsing with deionized water and squeezing the water out of the sponge pad 3-5 times.

Can I purchase replacement parts for the XCell SureLock Mini-Cell?

Here are the replacement parts we offer for the XCell SureLock Mini-Cell:
Replacement part - Cat. No.
Buffer Dam for XCell SureLock Mini-Cell and XCell II Blot Module - EI0012
L-Buffer Chamber for XCell SureLock Mini-Cell and XCell II Mini-Cell - EI0013
Platinum Wire for XCell SureLock Mini-Cell (12 inch x 2) - EI9022
XCell SureLock Lid - EI0010
XCell SureLock Wedge - EI0011
Buffer Core with Cathode Bar for XCell SureLock Mini-Cell - EI9015
Gel Knife - EI9010
Sponge Pad for Blotting (8) - EI9052

What are the components of the XCell SureLock Mini-Cell?

The XCell SureLock Mini-Cell includes the lid, gel tension wedge, lower buffer chamber, buffer core, buffer dam, and gel knife.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.