missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ XCell SureLock™ Lid
Description
Applications:
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| For Use With (Equipment) | XCell SureLock™ Mini |
| Gel Size | Mini |
| Product Line | XCell SureLock |
| Type | Lid |
| Quantity | 1 Each |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?