missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NS 2028

Product Code. 18025802
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18025802 50 mg 50mg
18014772 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18025802 Supplier Tocris Bioscience™ Supplier No. 4517/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent soluble guanylyl cyclase (sGC) inhibitor

Potent soluble guanylyl cyclase (sGC) inhibitor (Ki = 8 nM). Blocks sGC activity in murine cerebellum induced by S-nitroso-glutathione and NMDA (IC50 values are 17 and 20 nM respectively). Inhibits VEGF-induced cGMP accumulation; abolishes VEGF-induced migration in postcapillary venular endothelial cells (CVEC).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 204326-43-2
Molecular Formula C9H5BrN2O3

Specifications

Quantity 50 mg
Formula Weight 269.05
Purity >98%
Molecular Weight (g/mol) 269.05
Chemical Name or Material NS 2028
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.