missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NS 9283

Description
NS 9283 is a positive allosteric modulator of α4β2 receptor; increases the potency of Ach-evoked currents ∼60 fold without affecting the maximum efficacy (HEK293-hα4β2 cells). Reduces the rate of recovery from desensitization and slows the rate of deactivation. Displays a synergistic effect on the Emax when given with NS206 at 3α:2β receptors.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | NS 9283 |
| Purity | >99% |
| Molecular Weight (g/mol) | 248.24 |
| Inhibitors | NS 9283 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?