missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NSC 636819
GREENER_CHOICE

Product Code. 18704898
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18704898

Brand: Tocris Bioscience™ 5287/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KDM4A/KDM4B inhibitor

NSC 636819 is a KDM4A/KDM4B inhibitor; inhibits histone demethylase activity on H3K9me3 and induces apoptosis in LNCaP prostate cancer cells in vitro. Also negatively regulates androgen-responsive genes.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,5-Bis[(1E)-2-(3,4-dichlorophenyl)ethenyl]-2,4-dinitrobenzene
CAS 1618672-71-1
Quantity 10 mg
Target Histone Demethylase Inhibitors
Molecular Formula C22H12Cl4N2O4
Formula Weight 510.15
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.