missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals NSC 663284

Product Code. 15427919
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427919 10 mg 10mg
15417919 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427919 Supplier Thermo Scientific Chemicals Supplier No. J66520.MA
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 383907-43-5 | C15H16ClN3O3, C15H16ClN3O3 | 321.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 383907-43-5
Molecular Formula C15H16ClN3O3
Synonym 6-Chloro-7-(2-morpholin-4-yl-ethylamino)quinoline-5,8-dione; Cdc25 Phosphatase Inhibitor II

Specifications

Quantity 10 mg
Description Red
Content And Storage -30°C to -10°C
Formula Weight 321.76g/mol
Sensitivity Light Sensitive
Form Crystalline solid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.