missing translation for 'onlineSavingsMsg'
Learn More

NSC 91800, TRC

Product Code. 30424775
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424775 5 mg 5mg
30414518 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424775 Supplier Toronto Research Chemicals Supplier No. TRCN9004405MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13560-91-3
Molecular Formula C15H6Cl12
Molecular Weight (g/mol) 611.64
Synonym NSC 91800, 1,2,3,4,5,6,7,8,10,10,11,11-Dodecachloro-4,4a,4b,5,8,8a,9,9a-octahydro-1,4:5,8-dimethano-1H-fluorene
SMILES ClC1=C(Cl)C2(Cl)C3C(CC4C3C5(Cl)C(=C(Cl)C4(Cl)C5(Cl)Cl)Cl)C1(Cl)C2(Cl)Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H6Cl12/c16-6-8(18)12(22)4-2(10(6,20)14(12,24)25)1-3-5(4)13(23)9(19)7(17)11(3,21)15(13,26)27/h2-5H,1H2
Formula Weight 605.6732
Chemical Name or Material 1,2,3,4,5,6,7,8,10,10,11,11-Dodecachloro-4,4a,4b,5,8,8a,9,9a-octahydro-1,4:5,8-dimethano-1H-fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.