missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NTRK1/CEN1p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
1475.00 € - 2430.00 €
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | neurotrophic tyrosine kinase, receptor, type 1 |
Gene Alias | DKFZp781I14186,MTC,TRK,TRK1,TRKA,p140-TrkA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033325
|
Abnova™
FG0202.100UL |
100 μL |
1475.00 €
100µL |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043325
|
Abnova™
FG0202.200UL |
200 μL |
2430.00 €
200µL |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
DKFZp781I14186,MTC,TRK,TRK1,TRKA,p140-TrkA | |
4914 | |
Liquid |
Tube | |
neurotrophic tyrosine kinase, receptor, type 1 | |
NTRK1 | |
DNA/RNA |