missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies NU7026

Product Code. 17130342
Encompass_Preferred
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 17130342

Brand: STEMCELL Technologies 74174

This item is not returnable. View return policy

NHEJ pathway inhibitor; Inhibits DNA-dependent protein kinase (DNA-PK)

  • NU7026 is an inhibitor of DNA-dependent protein kinase (DNA-PK), an enzyme involved in the non-homologous end joining (NHEJ) DNA repair pathway
  • NU7026 sensitizes cells to radiation and has potential for anticancer therapies (Veuger et al.) NU7026 is also reported to increase the efficiency of homology-directed repair (HDR) in CRISPR-Cas9 genome editing (Riesenberg & Maricic; Zhang et al.)
  • Increases precise genome editing by promoting HDR at the expense of NHEJ in human pluripotent stem cells (Riesenberg & Maricic; Zhang et al.)
  • Sensitizes human cancer cell lines to DNA double-strand break-inducing therapy (chemo- or radio-therapy) by inhibiting DNA-PK activity and inducing cell cycle arrest at G2/M phase (Albarakati et al.; Ma et al.; Niazi et al.; Willmore et al.; Yang et al.)

Specifications

CAS 154447-35-5
Synonym LY293646; DNA-PK Inhibitor II
Quantity 25 mg
Molecular Formula C17H15NO3
Purity ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.