missing translation for 'onlineSavingsMsg'
Learn More

MP BIOMEDICALS Nuc-Off Nucleases and DNA Removal Spray

Product Code. 30214852 Shop All MP Biomedicals Products
Change view
Click to view available options
Volume (Metric):
450 mL
1800 mL
Unit Size:
1.8 L
450mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
30214852 1800 mL 1.8 L
30214849 450 mL 450mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30214852 Supplier MP BIOMEDICALS Supplier No. 112462451
  • ONLINE EXCLUSIVE PRICE
    1025.00 €
    969.15 € / 1.8 L
    Save 55.85 €
    5% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Non-toxic spray designed to remove nucleases, DNA, and RNA.

MP Biomedicals Nuc-Off Nucleases and Nucleic Acid Removal Spray is a ready-to-use solution for fast and cost-effective removal of RNase, DNAase, RNA and DNA.

This non-toxic spray-type solution can effectively remove nucleases, RNA and DNA from the surface of:

  • laboratory consumables
  • instruments
  • other utensils

creating a clean environment for RNA or DNA-related experiments.

It is strongly recommended to regularly clean your:

  • pipettors
  • bench surfaces
  • instrument exterior

Application Notes:

Nuc-Off Nucleases and Nucleic Acid Removal Spray is intended for effective and safe elimination of Nucleases, DNA, and RNA that may cause false positive or false negative experimental results.

Usage Statement:

Research Use Only (RUO)

Specifications

Product Type Removal Agent
Volume (Metric) 1800 mL
Product Identifier
  • Nuc-Off Nucleases and Nucleic Acid Removal Spray
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.