missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nuclear Fast Red, 94%, pure

Product Code. 10711745 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10711745 5 g Glass bottle 5g
10144351 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10711745 Supplier Thermo Scientific Chemicals Supplier No. 211980050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6409-77-4 | C14H11NNaO7S | 360.292 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6409-77-4
Molecular Formula C14H11NNaO7S
Molecular Weight (g/mol) 360.292
MDL Number MFCD00078493
InChI Key KDEATZKCHKVXGA-UHFFFAOYSA-N
Synonym Calcium Red, C.I. 60760
PubChem CID 131675027
IUPAC Name 4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium
SMILES [HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na]

Specifications

Melting Point >300.0°C
CAS Min % 92.0
CAS Max % 100.0
Assay Percent Range 94%
Solubility Information Solubility in water: 1g/L (25°C).
Formula Weight 357.26
Percent Purity 94%
Grade Pure
Packaging Glass bottle
Quantity 5 g
Chemical Name or Material Nuclear Fast Red
Show More Show Less
compliance-icons
Product Identifier
  • Nuclear Fast Red
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.