missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Nuclease Mix

Product Code. 10556115 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10556115 0.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10556115 Supplier Cytiva Supplier No. 80650142

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifically developed for removal of nucleic acids in sample preparation for IEF/2-D electrophoresis applications

Removal of nucleic acids is often required to avoid contamination and subsequent artifacts on 2-D gels. The Nuclease Mix offers an effective mix of DNase and RNase enzymes, as well as the necessary cofactors for optimal nuclease activity.

  • Compatible with Cytiva Protease Inhibitor Mix
  • Ready to use: add mix to sample and incubate
  • Does not contain EDTA, a known inhibitor of nucleases

Proteomics

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Removal of Nucleic Acids in sample preparation for IEF/2-D electrophoresis applications in proteomic studies
Content And Storage -20°C
Quantity 0.5 mL
Product Type Nuclease Mix
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.