missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nucleic Acid Detection Blocking Buffer

Product Code. 11820104 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11820104 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11820104 Supplier Thermo Scientific™ Supplier No. 89880A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Nucleic Acid Detection Blocking Buffer is intended for use with the Thermo Scientific Chemiluminescent Nucleic Acid Detection Module Kit, of which it is also a component.

Nucleic Acid Detection Blocking Buffer is intended for use with the Thermo Scientific Chemiluminescent Nucleic Acid Detection Module Kit, of which it is also a component. Non-specific binding of probe to membrane can be minimized by incubating the membrane in Blocking Buffer for 15 minutes.

Related Products

  • Chemiluminescent Nucleic Acid Detection Module Kit (Cat. No. 89880)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C.
Buffer Blocking Buffer
Product Type Nucleic Acid Detection Blocking Buffer
Quantity 500 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.