missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ PC EF, kit for endotoxin-free plasmid DNA
Click to view available options
No. of Reactions:
5
10
Yield:
100–500μg
2,000–10,000μg
500–2,000μg
Sample Volume:
30 - 150mL
150 - 500mL
500 - 2000mL
Test Time:
100 min./2 preps
150 min./2 preps
180 min./2 preps
Unit Size:
10 reactions
5 preps
5 reactions
Description
An important prerequisite for transfections and gene therapy applications is endotoxin-free plasmid DNA. The patented procedure of NucleoBond™ PC EF reduces endotoxins to a very low level using the same working procedure as the standard protocol. In comparison to competitive kits, there is no need for a time-consuming incubation step to remove the endotoxins.
Features
- Fast and easy clarification by NucleoBond™ Folded Filters—No centrifugation required, no shearing forces
- Proven anion-exchange technology—Yields transfection-grade high-copy plasmid DNA from E. coli
- NucleoBond™ Columns contain NucleoBond™ AX Silica Resin packed between inert filters
- Polypropylene columns are resistant to organic solvents and are free of DNase and RNase
- AX Resin can be used over a wide pH range and can remain in contact with buffers for up to three hours
Specifications
- DNA with less than 0.1 EU/μg DNA
- Purity of plasmid DNA: A260/A280 = 1.80–1.95
- Recovery of plasmid DNA >90%
- Structural integrity >90%
- Yield between 500μg up to 10mg, depending on kit used
Specifications
Specifications
| Concentration | Endotoxin level: <0.1 EU/μg |
| Format | Maxi Gravity-flow Column |
| Isolation Technology | Anion Exchange Chromatography Technology |
| Efficiency | Recovery of plasmid DNA: >90% |
| For Use With (Application) | High-copy plasmids from E. coli |
| Includes | NucleoBond AX 500 Columns, buffers, RNase A, NucleoBond Folded Filters XL |
| No. of Reactions | 10 |
| Product Type | NucleoBond™ PC 500 EF |
| Quantity | 1 Kit |
| Purity | A260/A280: 1.80–1.95 |
| Show More |
Safety and Handling
Hazard Category
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Respiratory sensitiser Category 1
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction