missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Midi Columns for transfection-grade plasmid DNA

Product Code. 15749999
Click to view available options
For Use With (Equipment):
NucleoBond Xtra Maxi
NucleoBond Xtra Midi
Unit Size:
Pack of 10

Product Code. 15749999

Brand: Macherey-Nagel™ 740414.10S

Please to purchase this item. Need a web account? Register with us today!

Edge

Polypropylene gravity flow columns with inserted column filters for use with NucleoBond Xtra Midi or Maxi kits for ultra-fast isolation of transfection-grade plasmid DNA with high yield.

NucleoBond™ Xtra—The new generation of anion exchanger for ultra-fast transfection-grade plasmid DNA

  • Optimized column design – Midi prep in approximately 30 min, Maxi prep in 35 min
  • Available in Midi and Maxi sizes – typical yields of 400 μg (Midi) and 1000 μg (Maxi)
  • Polypropylene columns suitable for use with organic solvents and buffers with denaturing agents
  • Column filter included for high filter flow rates, parallel lysate clearing, and loading onto the column
    • Depth filters avoid time-consuming centrifugation for lysate clearing
    • Complete removal of precipitate even with large lysate volumes without clogging
    • Avoids shearing of large DNA constructs (such as PACs or BACs)

Specifications

Content And Storage Room temperature (18°C to 25°C)
Isolation Technology Anion Exchange Chromatography Technology
Final Product Type Transfection grade DNA
For Use With (Equipment) NucleoBond Xtra Maxi
For Use With (Application) Plasmid DNA purification
Includes NucleoBond Xtra Column Filters
Product Type Gravity Flow Column
Quantity 10 Pk.
Purity A260/A280: 1.80 - 1.95
Sample Volume < 600 mL (high copy), < 1200 mL (low copy)
Size < 300 kbp
Test Time 35 min.
Yield 1000 μg
Show More Show Less
Product Identifier
  • NucleoBond Xtra Maxi
Supplemental information
  • MIXTURE LIST-Contains : 2-propanol/sodium hydroxide solution (diluted < 2 %),dodecylsulfate, sodium salt/ethanol (diluted <
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.