missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Combi Rack

Product Code. 11922572
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each

Product Code. 11922572

Brand: Macherey-Nagel™ 740415

Please to purchase this item. Need a web account? Register with us today!

Edge

Rack for use with NucleoBond Xtra Midi / Maxi, Xtra BAC, AX 100, AX 500, AX 2000, and AX 10000 Columns.

NucleoBond™ Xtra is a new generation of anion exchangers featuring fast filtration, high binding capacity and high flow rate. They yield ultra-pure plasmid DNA in about half the time compared with other Midi or Maxi kits based on anion-exchange chromatography. NucleoBond™ Xtra Midi and Maxi kits contain enlarged columns creating shorter silica resin beds.
  • Holds up to six (6) NucleoBond™ Xtra Midi and four (4) NucleoBond™ Xtra Maxi Columns
The Xtra Combi Rack holds columns from either kit; it is sold separately.

Specifications

Content And Storage 1 Ea., 15°C to 25°C
Description Holds up to six (6) NucleoBond™ Xtra Midi and four (4) NucleoBond™ Xtra Maxi Columns
Final Product Type Nucleo Bond Xtra Combi Rack
For Use With (Equipment) NucleoBond Xtra Midi, Maxi, BAC, AX 100, 500, 2000, And 10000, BAC 100 Columns
Product Type Nucleo Bond Xtra Combi Rack
Quantity 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.