missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Maxi EF, Maxi kit for endotoxin-free plasmid DNA

Product Code. 11982482
Change view
Click to view available options
No. of Reactions:
10
50
Unit Size:
10 reactions
2.75 L
50 preps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
11982482 10 10 reactions
12728422 10 2.75 L
12738422 50 50 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11982482 Supplier Macherey-Nagel™ Supplier No. 740424.10
  • ONLINE EXCLUSIVE PRICE
    500.25 €
    386.40 € / 10 reactions
    Save 113.85 €
    22% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

2nd generation of anion exchanger for time saving endotoxin-free plasmid DNA. Purify endotoxin-free plasmid DNA with high yield. The NucleoBond™ Xtra Maxi EF Kit represents the new generation of anion exchanger allowing high yield, ultra-fast.

Features
  • Endotoxin-free
  • Plasmid DNA with less than 0.05 EU/μg for transfection of highly sensitive cells
  • Patented endotoxin removal by additional washing step – no extra ice incubation necessary
  • Optimized column design – Maxi prep in 50 min.
  • Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
  • Improved silica material – typical yields of 1000μg (Maxi)
Applications
  • High-copy plasmids from E. coli
  • Low-copy plasmids from E. coli (increased buffer volumes required)
  • High-speed Version

Specifications

Isolation Technology Silica Membrane Technology
Formulation Endotoxin-free Plasmid Maxi preps
No. of Reactions 10
Sample Volume <600 mL (high copy), <1200 mL (low copy)
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoBond Xtra Maxi EF
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: 2-propanol/ethanol (diluted < 20 %)/Sodium hydroxide solution (diluted < 2 %), dodecyl sulfate, sodi
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.