missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra Midi EF, Midi kit for endotoxin-free plasmid DNA

Product Code. 12773550
Change view
Click to view available options
No. of Reactions:
10
50
Unit Size:
10 reactions
50 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
12773550 10 10 reactions
12798412 50 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12773550 Supplier Macherey-Nagel™ Supplier No. 740420.10
  • ONLINE EXCLUSIVE PRICE
    213.17 €
    169.05 € / 10 reactions
    Save 44.12 €
    20% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Purify endotoxin-free plasmid DNA with high yield. The NucleoBond™ Xtra Midi EF Kit represents the new generation of anion exchanger allowing high yield, ultra-fast.

Features
  • Endotoxin-free
  • Plasmid DNA with less than 0.05 EU/μg for transfection of highly sensitive cells
  • Patented endotoxin removal by additional washing step – no extra ice incubation necessary
  • Optimized column design – Midi prep in 45 min.
  • Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
  • Improved silica material – typical yields of 400μg (Midi)
Applications
  • High-copy plasmids from E. coli
  • Low-copy plasmids from E. coli (increased buffer volumes required)

Specifications

Buffer Yes
Format Midi Gravity-flow Column
Isolation Technology Anion Exchange Chromatography Technology
No. of Reactions 10
RNase Yes
Sample Volume <200 mL (high copy), <400 mL (low copy)
Size <300 kbp
Yield 400μg
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoBond Xtra Midi EF
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: 2-propanol/ethanol(diluted < 20 %)/sodium hydroxide solution(diluted < 2 %),dodecyl sulfate,sodium s
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.