missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ Xtra Midi Plus kit for transfection-grade plasmid DNA
Description
Features
- Proven anion-exchange technology – transfection-grade plasmid DNA
- Patented endotoxin removal by additional washing step – no extra ice incubation necessary
- Optimized column design – Midi prep in 30 min.
- Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
- Improved silica material – typical yields of 400μg (Midi)
- NucleoBond™ Xtra EF kits contain NucleoBond™ Finalizers to speed up DNA precipitation
- High-copy plasmids from E. coli
- Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications
Specifications
| Buffer | Yes |
| Format | Midi Gravity-flow Column |
| Isolation Technology | Anion Exchange Chromatography Technology |
| No. of Reactions | 10 |
| RNase | Yes |
| Sample Volume | <200 mL (high copy), <400 mL (low copy) |
| Size | <300 kbp |
| Yield | 400μg |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: 2-propanol/sodium hydroxide solution (diluted < 2 %),dodecylsulfate, sodium salt/ethanol (diluted <
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?