missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag Tissue and Trace kit for DNA purification

Product Code. 12788662
Change view
Click to view available options
No. of Reactions:
1 x 96
24 x 96
4 x 96
Unit Size:
2304 reactions
384 reactions
96 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
12788662 4 x 96 384 reactions
12798672 1 x 96 96 reactions
12778662 24 x 96 2304 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12788662 Supplier Macherey-Nagel™ Supplier No. 744300.4
  • ONLINE EXCLUSIVE PRICE
    1235.00 €
    1094.57 € / 384 reactions
    Save 140.43 €
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flexible magnetic bead based purification of DNA from a broad range of samples and flexible magnetic bead based isolation of DNA from forensic traces.

NucleoMag 96 Tissue is designed for the rapid parallel purification of genomic DNA from animal tissues, cell cultures and bacteria in a 96 well format. NucleoMag 96 Trace is designed for the purification of DNA from forensic samples (e.g. buccal swabs, dried blood spots, cigarette filters). NucleoMag 96 Plant is designed for the purification of genomic DNA from plant tissues in a 96 well format.

The magnetic bead technology provides a closed system therefore, the risk for cross contamination is greatly reduced.The NucleoMag technology can be easily adapted to common laboratory automation platforms. It can be used either with static-pin separators (eg NucleoMag SEP) or magnetic separators integrated into robotic workstations.

Kit components:

  • NucleoMag 96 Tissue – NucleoMag B-beads, buffers, Proteinase K
  • NucleoMag 96 Trace – NucleoMag B-beads, buffers, Proteinase K
  • NucleoMag 96 Plant – NucleoMag C-beads, buffers.

Rapid, economic purification of genomic DNA using highly reactive superparamagnetic beads, easily adapted to automated use.

  • Sample size: NucleoMag 96 Tissue – up to 20mg animal tissue, 1 x 107 cells or bacteria NucleoMag 96 Trace – buccal swabs, dried blood spots, cigarette filters NucleoMag 96 Plant – 20mg to 50 mg plant tissue
  • Typical yield: NucleoMag 96 Tissue/Plant – 10μg to 20μg DNA NucleoMag 96 Trace – 1μg to 3μg DNA
  • A260/280 ≥1.6 to 1.9
  • Concentration of sample possible
  • Complete removal of PCR* inhibitors
  • Small elution volumes (≥50μL)
  • Yield does not depend on elution volume
  • DNA ready to use in downstream applications
  • Binding capacity: 0.4μg DNA per μL of NucleoMag bead suspension
  • Size of beads: 1μm to 3μm
  • Reduced carry-over of beads

For the manual and automated purification of gDNA from a wide range of sources.

Specifications

Capacity 0.4 μg/μL beads
Isolation Technology Magnetic Bead
For Use With (Application) For Manual and Automated Isolation of DNA
No. of Reactions 4 x 96
Product Type NucleoMag™ 96 B-Bead
Sample Volume <20 mg human/animal tissue, <106 cells
Test Time 120min.
Yield 10 to 20μg
Form Highly Reactive Superparamagnetic Beads
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoMag 96 Tissue
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash with water thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: ethanol (denatured with 1%IPA/1%MEK, acc.2016/1867/EU), sodium perchlorate, proteinase K (origin: tr
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.