missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag Plant kit for DNA purification from plants

Product Code. 12728672
Change view
Click to view available options
No. of Reactions:
24 x 96
4 x 96
Unit Size:
2304 reactions
384 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
12728672 4 x 96 384 reactions
12795224 24 x 96 2304 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12728672 Supplier Macherey-Nagel™ Supplier No. 744400.4
  • ONLINE EXCLUSIVE PRICE
    823.00 €
    717.27 € / 384 reactions
    Save 105.73 €
    12% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flexible magnetic bead based isolation of DNA from a multitude of plant samples.

After lysis of the blood cells genomic DNA is selectively bound to the NucleoMag Blood beads. Contaminants are washed away and the DNA can be eluted from the beads under low-salt conditions.As the magnetic bead technology provides a closed system (no transfer of sample from one well to another is necessary during the preparation) the risk for cross-contamination is greatly reduced. The NucleoMag technology can be easily adapted to common laboratory automation platforms. It can be used either with static-pin separators (eg. NucleoMag SEP) or magnetic separators integrated into robotic workstations.Kit components: NucleoMag B-Beads, elution plate (u-bottom), buffers. For further information on this product please contact our Technical Support team.
  • Magnetic bead technology
  • Yield: 1μg to 4μg
  • Sample material: 100μL whole blood
  • Elution volume: 25μL to 100μL
  • Binding capacity: 0.4μL/1μL beads
  • Preparation time: 120 min./96 preps
  • Fragment size: 300bp to 30kb
  • Format: superparamagnetic beads (size of beads: 1μm to 3μm)
For manual or automated isolation of genomic DNA from whole blood.

Specifications

Capacity 0.4 μg/μL beads
Isolation Technology Magnetic Bead
For Use With (Application) For Manual and Automated Isolation of DNA
No. of Reactions 4 x 96
Product Type NucleoMag™ 96 C-Bead
Test Time 120min.
Yield 10 to 20μg
Form Highly Reactive Superparamagnetic Beads
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoMag 96 Plant
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Respiratory sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: dodecyl sulfate, sodium salt / 2-propanol / ethanol, sodiumperchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.