missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag DNA Swab for DNA purification from swabs

Product Code. 16322028
Change view
Click to view available options
No. of Reactions:
1 x 96
24 x 96
4 x 96
Unit Size:
192 reactions
2304 reactions
96 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
16322028 1 x 96 96 reactions
16332028 4 x 96 192 reactions
15880978 24 x 96 2304 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
UN 1993 Flammable liquid, n.o.s. (ethanol mixture) Chem class 3 Product group II
Product Code. 16322028 Supplier Macherey-Nagel™ Supplier No. 744601.1
  • ONLINE EXCLUSIVE PRICE
    337.00 €
    299.02 € / 96 reactions
    Save 37.98 €
    11% Off
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
UN 1993 Flammable liquid, n.o.s. (ethanol mixture) Chem class 3 Product group II

Magnetic bead based kit for the isolation of genomic DNA from swabs.

  • High throughput DNA isolation for genetic testing
  • Developed for cotton as well as synthetic swabs
  • Combine with NucleoSpin™ Forensic Filters for convenient sample prep

Specifications

Capacity 0.4 μg/μL beads
Concentration 10 to 30 ng/μL
Isolation Technology Magnetic Bead
Sample Type Swab (Cotton or Synthetic)
Efficiency 0.4 μg/μL beads theoretical binding capacity
For Use With (Equipment) COPAN FLOQSwabs™, Puritan HydraFlock™ swabs
For Use With (Application) Typical downstream applications: qPCR, NGS, STR analysis, any kind of enzymatic reaction
No. of Reactions 1 x 96
Purity A260/A280: 1.6 - 1.9
Test Time 30 min/ 96 preps on KingFisher™ Flex (excluding lysis)
Form Highly Reactive Superparamagnetic Beads
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • NucleoMag DNA Swab (1x96)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
Supplemental information
  • MIXTURE LIST-Contain: guanidium hydrochloride, proteinase k, glycerole, ethanol

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.