missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag kit for clean up and size selection of NGS library prep reactions

Product Code. 15899167
Click to view available options
Quantity:
5 mL
50 mL
500 mL
Unit Size:
500mL
50mL
5mL
This item is not returnable. View return policy

Product Code. 15899167

Brand: Macherey-Nagel™ 744970.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NucleoMag™ NGS Beads Suspension that enable magnetic bead-based clean-up and size selection of NGS library preparation reactions for use in Next Gen Sequencing.

NucleoMag™ NGS Clean-up and Size Select

  • Clean-up and size selection steps in NGS library kits
  • Sample input as little as 17.5 pg
  • Sample volume of 50 μL to 150 μL
  • Tunable size selection 150-800 bp
  • Convenient magnetic bead technology
  • Easy to adjust for specific applications or sequencers
  • Optimal scalability for manual and automated processing

Specifications

Content And Storage Refrigerated at 4 - 8 °C
Isolation Technology Magnetic Bead
Sample Type Reaction Mixtures from Common NGS Library Kits
Elution Volume Elution volume: 10 - 100 μL
For Use With (Equipment) Magnetic separator; Elution and separation plates
For Use With (Application) NGS clean-up and size selection
Includes NucleoMag NGS-Beads in buffer
Quantity 50 mL
Sample Volume 17.5 pg to 5 μg, Input volume: 50 - 150μL
Yield Typical recovery: ≥ 80 %
Form Highly Reactive Superparamagnetic Beads
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.