missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoMag RNA kit for magnetic bead based RNA purification
Click to view available options
No. of Reactions:
4 x 96
96
Unit Size:
384 reactions
96 reactions
Description
Features
- One-tube procedure minimizes risk of cross-contamination
- Small elution volumes ≥ 50μL
- Suitable for manual and automated processing
- Recombinant Dnase included
- Reducing agent TCEP included (no β-mercaptoethanol necessary)
- Rapid manual and automated small-scale preparation of highly pure total RNA from tissue or cell samples
Specifications
Specifications
| Format | Highly Reactive Superparamagnetic Beads |
| For Use With (Application) | Rapid manual and automated small-scale preparation of highly pure total RNA from tissue or cell samples |
| Includes | NucleoMag™ B-Beads, buffers, TCEP, RNase-free rDNase |
| No. of Reactions | 4 x 96 |
| Product Type | NucleoMag RNA |
| RNase | Yes |
| Sample Size | <2 x 106 cells, <20 mg human/animal tissue |
| Test Time | <120 min./96 Reaction |
| Yield | <30μg |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 2
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- LONG-TERM AQUATIC HAZARD Chronic 4
- H225-Highly flammable liquid and vapour.
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- H413-May cause long lasting harmful effects to aquatic life.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : guanidinium thiocyanaten,ethanol/tris-(2-carboxyethyl)phosphine hydrochloride, TCEP(
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction