missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoProtect RNA stabilization reagent for cells and tissues.
Click to view available options
Quantity:
50 mL
250 mL
500 mL
Unit Size:
250mL
500mL
50mL
Description
- Collect the sample first, get the RNA later
- Protect your cell and tissue samples from RNA degradation
- Compatible with most RNA isolation methods, includes NucleoSpin and NucleoMagRNA kits as well as NucleoZOL
Specifications
Specifications
| Content And Storage | ≤ 24 hours at 37°C, ≤ 7 days at 18 to 25°C, ≤ 1 month at 4°C, long term at -20°C or lower |
| Sample Type | Human/Animal Cells/Tissues (Max. 5 mm dia.) |
| For Use With (Equipment) | Safe, transport of stabilized cell and tissue samples without dry ice for subsequent RNA isolation |
| For Use With (Application) | Preservation of RNA integrity within cell and tissue samples (downstream applications: qRT-PCR, RNAseq,array technology, etc.) |
| Purity | Typical RIN after RNA isolation: 10 for cultured mammalian cells (e.g., HeLa); > 9 for mammalian |
| Quantity | 500 mL |
| Form | RNA Stabilization Reagent for Cells and Tissues |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction