missing translation for 'onlineSavingsMsg'
Learn More

Nucleoside-Analog-2, MedChemExpress

Product Code. 16420678
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16420678 5 mg 5mg
16414127 50 mg 50mg
16408238 10 mg 10mg
16407966 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16420678 Supplier Medchem Express Supplier No. HY77652
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nucleoside-Analog-2 is a 4'-Azidocytidine analogue against Hepatitis C virus (HCV) replication.

Nucleoside-Analog-2 is a 4'-Azidocytidine analogue against Hepatitis C virus (HCV) replication.

Specifications

Chemical Name or Material Nucleoside-Analog-2
CAS 876708-01-9
Color Gray
Purity Grade Notes Research
Molecular Formula C9H11N5O6
Quantity 5 mg
Solubility Information DMSO : 33.33 mg/mL (116.86 mM; Need ultrasonic) ∣H2O : 20 mg/mL (70.12 mM; Need ultrasonic)
SMILES O[C@@H]1[C@](CO)(N=[N+]=[N-])O[C@@H](N2C(NC(C=C2)=O)=O)[C@H]1O
Molecular Weight (g/mol) 285.21
Formula Weight 285.21
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.