missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSnap cfDNA kit for cell-free DNA from plasma
Click to view available options
No. of Reactions:
10
50
Dimensione della confezione:
10 reactions
50 reactions
Descrizione
The NucleoVac 24 Vacuum Manifold allows the processing of silica columns for nucleic acid purification by vacuum.
- Snap-off column design for quick processing of large sample volumes
- Compatible with the NucleoVac 24 vacuum manifold (sold separately, MPN 740299
- No need for carrier-RNA
- Process: DNA precipitation—filtration—washing—elution
- Optimized protocol for use with EDTA or Cell-Free DNA BCT™ (Streck) blood draw tubes
- Highly efficient recovery of cell-free DNA from Liquid Biopsies
- No alcohol precipitation necessary
- Typical downstream applications: NGS, qPCR, ddPCR
Specifica
Specifica
| Format | Snap-off Column |
| Isolation Technology | Silica Membrane Technology |
| Sample Type | Plasma, Urine |
| Elution Volume | Elution volume: 20 - 100 μL |
| Final Product Type | Cell-free DNA |
| For Use With (Equipment) | Vacuum (e.g. NucleoVac 24 Vacuum Manifold and vacuum pump) |
| For Use With (Application) | DNA isolation from blood plasma |
| Includes | NucleoSnap DNA Plasma Columns, Collection Tubes, buffers, Proteinase K |
| No. of Reactions | 50 |
| Purity | Fragment size: ≥ 50 bp |
| Vedi altri risultati |
Sicurezza e movimentazione
Identificativo prodotto
- NucleoSnap DNA Plasma
- Danger
- Flammable liquid Category 2
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- MIXTURE LIST-Contains : potassium hydroxidesolution/proteinaseK, liquid (origin: tritirachium album)/guanidine hydrochlori
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto