missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 8 RNA, 8-well kit for RNA purification

Product Code. 12712613
Change view
Click to view available options
Sample Size:
<2 x 106 cultured cells, <20 mg human/animal tissue
Unit Size:
480 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Size Unit Size
12712613 <2 x 106 cultured cells, <20 mg human/animal tissue 480 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12712613 Supplier Macherey-Nagel™ Supplier No. 740698.5
  • ONLINE EXCLUSIVE PRICE
    2155.00 €
    1946.94 € / 480 reactions
    Save 208.06 €
    9% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

RNA isolation in flexible 8‐well strip format.

NucleoSpin 8/96 RNA for automated purification of total RNA from animal or human cell cultures and tissue combines the stringency of guanidine-isothiocyanate lysis with effective digestion of genomic DNA by DNase I treatment and the speed of silica-membrane technology. NucleoSpin 8/96 RNA means consistent RNA quality, reproducible yields of RNA, and high-throughput procedures free of cross-contamination. Therefore, NucleoSpin 8/96 RNA kits are powerful products for e.g. microarray analysis in respect to gene expression profiling, drug discovery, and molecular diagnostics (see Application data). The kits allow purification of total RNA from up to 107 cells or up to 30mg tissue (centrifuge processing). The unique 8-well strip/96-well plate design of the NucleoSpin 8/96 RNA systems allow the fully automated drying of the RNA binding module before loading of elution buffer and minimize spraying. With the NucleoSpin 8/96 RNA method, cells or tissue are lysed by incubation in a solution containing large amounts of chaotropic ions (cells can be grown and directly lysed in 96-well cell culture plates). This lysis buffer immediately inactivates RNases and creates appropriate binding conditions which favour adsorption of RNA to the silica membrane of the NucleoSpin 8 RNA Binding Strip/96 RNA Binding Plate.Contaminating DNA, which is also bound to the silica membrane, is removed by DNase I which is directly applied onto the membrane during the preparation (DNase I is supplied with the kits). Due to the high specific activity of the DNase I a treatment of only 15min at room temperature is sufficient for an efficient removal of contaminating DNA. Salts, metabolites, and macromolecular cellular components are removed by simple washing steps with three different buffers. Pure RNA is finally eluted under low ionic strength conditions with RNase-free water (pH 8.0 to 8.5).

Kit components:

NucleoSpin 8 RNA kit - NucleoSpin RNA Binding Strips, MN Wash Plates, MN Square-well Blocks, Racks with Tube Strips, Cap Strips, Elution plates U-bottom, Self-adhering Foil, buffers, RNase-free DNase INucleoSpin 96 RNA kit - NucleoSpin RNA Binding Plates, MN Wash Plates, Square-well Blocks, Round-well Block (low), Elution Plates U-bottom, Self-adhering Foil, buffers, RNase-free DNase I

Manual or automated isolation of total RNA from cultured cells and tissue, saliva samples collected with Oragene.

Specifications

Includes NucleoSpin RNA Binding Strips, MN Wash Plates, MN Square-well Blocks, Racks with Tube Strips, Cap Strips, Elution plates U-bottom, Self-adhering Foil, buffers, RNase-free DNase I
No. of Reactions 60 x 8
Product Type NucleoSpin 8 RNA
Sample Size <2 x 106 cultured cells, <20 mg human/animal tissue
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoSpin 8 RNA (60x8)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: guanidine thiocyanate, rdnase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.