missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ 96 Blood, 96-well kit for DNA from blood
Click to view available options
:
No. of Reactions: 1 x 96
Description
NucleoSpin™ 96 Blood is designed for the parallel isolation of genomic DNA from whole blood. After lysis of blood cells genomic DNA is selectively bound to a silica membrane while contaminants are washed away effectively. Elution is possible even under vacuum without cross-contamination. Due to its plate design and the possibility to perform the complete procedure under vacuum NucleoSpin™ 96 Blood can be easily adapted to common liquid handling instruments for fully automated processing without manual interactions. Kit components: NucleoSpin™ Blood binding plate, MN wash plate, buffers, proteinase K, 15mL tubes, cap strips, square-well block, rack with tube strips.
Required hardware:
Vacuum processing - the NucleoSpin 96 blood kits can be used manually with the NucleoVac 96 vacuum manifold. For vacuum processing of the 8-well strips the reusable starter set A, containing column holders A and dummy strips is required. For automation on laboratory platforms with standard 96-well vacuum chambers the use of the starter set A is also required.- Silica membrane technology
- Yield: 4μg to 6μg
- Sample material: ≤200uL whole blood/2 x 106 cultured cells
- Elution volume: 100μL
- Binding capacity: 20μg
- Preparation time: 70 min./plate
- Format: 96 well plates
- Fragment size: 300bp to 30kb
- No use of organic solvents Suitable for whole blood (human or animal); Whole blood treated with citrate, EDTA, heparin.
Specifications
Specifications
| No. of Reactions | 1 x 96 Preps |
| Product Type | 96 Blood |
Safety and Handling
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction