missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 Plasmid, 96-well kit for plasmid DNA

Product Code. 12857321
Click to view available options
No. of Reactions:
24 x 96 Preps
4 x 96 preps
Unit Size:
2304 reactions
Pack of 4
This item is not returnable. View return policy

Product Code. 12857321

Brand: Macherey-Nagel™ 740625.24

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for manual or automated large-scale purification of high-copy plasmid DNA from E.coli in the 96-well plate format. Macherey-Nagel™ NucleoSpin™ 96 Pasmid Kit is an ultra-fast and easy-to-use kit that supplies all core and accessory plates, buffers and RNase A for the highest convenience.

NucleoSpin™ 96 Pasmid kits enable the high-throughput isolation of sequencing-grade plasmid DNA from E. coli. The proven 96-well plate format allows for easy automation on common liquid handling instruments or for manual processing. Up to 96 samples can be processed simultaneously, typically within less than 45 minutes. Uses include isolation of high-copy plasmids, purification of cosmids and reliable downstream applications, such as cloning, sequencing, PCR, transformation, and restriction analysis.

  • Time-saving parallel isolation of plasmid DNA
  • Processing of up to 5mL bacterial culture
  • 96-Well filter plate for convenient filtration of bacterial lysates
  • Suitable for manual or automated processing under vacuum or by centrifugation
  • Innovative MN Wash Plate minimizes risk of cross-contamination
  • A260/A280: 1.70–1.85; Binding capacity: 20μg

Specifications

Format 96-well Plate
Sample Type E. coli Culture
Elution Volume Elution volume: 75–150μL
Final Product Type Sequencing grade plasmid DNA
For Use With (Equipment) Liquid handling instruments
For Use With (Application) Manual and automated processing under vacuum or by centrifugation
Includes NucleoSpin Plasmid Filter Plate (24), NucleoSpin Plasmid Binding Plate (24), MN Wash Plate (24), Culture plate with Gas-permeable Foil (24), Elution Plate with Self-adhering Foil (24), RNase A (6x60mg), Resuspension Buffer A1 (6x150mL), Lysis Buffer A2 (6x150mL), Neutralization Buffer A3 (6x200mL), Wash Buffer AW (6x400mL), Wash Buffer A4 Concentrate (6x200mL), Elution Buffer AE (6x125mL)
No. of Reactions 24 x 96 Preps
Product Type NucleoSpin™ 96 Plasmid Kit
Quantity 1 Kit
Sample Volume 1–5mL
Test Time Preparation time: 45 min. / plate
Yield 4–6μg plasmid per 1mL E. coli culture
Maximum plasmid size: <15 kbp
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide solution (diluted < 2 %), dodecyl sulfate,sodium salt / guanidine hydrochloride, 2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.