Learn More
Macherey-Nagel™ NucleoSpin™ 96 Plasmid Transfection-grade Core Kit, 96-well kit
Description
Endotoxins are co-purified during typical plasmid preparations from bacterial lysates. Since they interfere with eukaryotic cell survival, endotoxin reduction is essential prior to cell transfection. NucleoSpin™ Plasmid Transfection-grade kits are designed to reduce endotoxins from bacterial lysates.
- Purifies high-copy plasmids for transfection
- Novel technology to diminish endotoxin content (< 50 EU/μg DNA) for successful transfection
- Rapid purification from E. coli via automatic, manual, vacuum or centrifugation processing
- Ultra-fast procedure simplifies the work-flow
- High-throughput — 96-well plates allow for easy automation and supports plasmid screening
- Downstream applications: transfection of common cell lines (lipofection, electroporation)
- Also suitable for standard molecular biology application, such as cloning and sequencing
Spécification
Spécification
| Bacterial Endotoxins | <50 EU/ng DNA |
| Content And Storage | Room temperature (18 - 25 °C) |
| Format | 96-well Plate |
| Isolation Technology | Silica Membrane and Endotoxin-reduction Technology |
| Sample Type | Bacterial Culture (E. coli, High-copy Plasmids) |
| Efficiency | Binding capacity: 20 μg |
| Elution Volume | Elution Volume: 100 - 200 μL |
| Final Product Type | Transfection-grade plasmid DNA |
| For Use With (Equipment) | Centrifuge, Vacuum manifold (e.g. NucleoVac 96) |
| For Use With (Application) | Rapid purification of plasmids for transfection |
| Afficher plus |
Sécurité et traitement
- NucleoSpin 96 Plasmid T.-grade (4x96)
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: sodium hydroxide, dodecyl sulphate, guanidine hydrochloride,triethanolamine, 2-propanol, RNAse
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.