missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 RNA Plates for RNA purification

Product Code. 15830948
Change view
Click to view available options
Quantity:
4 Plates
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15830948 4 Plates Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15830948 Supplier Macherey-Nagel™ Supplier No. 740709.4S
  • ONLINE EXCLUSIVE PRICE
    797.00 €
    672.00 € / Pack of 4
    Save 125.00 €
    15% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

RNA isolation in proven 96‐well plate format

NucleoSpin™ 96 RNA Kit/Core Kit

  • Time-saving parallel isolation of total RNA
  • Proven 96-well format for high throughput
  • DNase included for efficient removal of genomic DNA
  • Processing under vacuum or by centrifugation
  • Suitable for manual and automated processing
  • Innovative MN Wash Plate minimizes risk of cross-contamination
  • RNA ready to use for any kind of enzymatic reaction
  • Core Kits include basic content focussed on automation platforms

Specifications

Color Light blue band
Content And Storage Room temperature (18 - 25 °C)
Format 96-well Plate
Isolation Technology Silica Membrane
Elution Volume 50 - 130 μL
Sample Type Cultured Cells, Saliva, Tissue
Efficiency Binding capacity: 100 μg
Final Product Type Total RNA
For Use With (Equipment) NucleoSpin 96 RNA kit or core kit
For Use With (Application) Isolation of RNA
Product Type Binding Plate
Quantity 4 Plates
Sample Size <2 x 106 cultured cells, <20 mg human/animal tissue
Test Time 70 min/plate
Show More Show Less
Product Identifier
  • NucleoSpin 96 RNA
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate/ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.