missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 RNA, 96-well kit for RNA purification

Product Code. 12768542
Change view
Click to view available options
No. of Reactions:
2 x 96
24 x 96
4 x 96
Unit Size:
192 reactions
2304 reactions
384 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
12768542 24 x 96 2304 reactions
12736543 2 x 96 192 reactions
12756543 4 x 96 384 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12768542 Supplier Macherey-Nagel™ Supplier No. 740709.24
  • ONLINE EXCLUSIVE PRICE
    9875.00 €
    8473.50 € / 2304 reactions
    Save 1401.50 €
    14% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for manual or automated high throughput isolation of RNA from animal or human tissue. Macherey-Nagel™ NucleoSpin™ 96 RNA Kit enables time-saving parallel isolation of total RNA. The kit supplies all core content and convenient accessories for the highest efficiency.

NucleoSpin™ 96 RNA kits enable the isolation of RNA from up to 96 samples of fresh, frozen or stabilized animal or human cell cultures. NucleoSpin 96 RNA delivers consistent RNA quality, reproducible yields of RNA, and high-throughput procedures free of cross-contamination. The kits combine the stringency of guanidine-isothiocyanate lysis with effective digestion of genomic DNA by DNase I treatment and the speed of silica-membrane technology.

  • Time-saving parallel isolation of RNA from cultured cells or from saliva samples collected with Oragene™ RNA.
  • Suitable for manual processing under vacuum or by centrifugation, or used fully automated on common laboratory workstations.
  • Includes rDNase for efficient removal of genomic DNA and innovative MN Wash Plate to minimize the risk of cross-contamination.
  • Produces the highest purity total RNA, suitable for direct use in any kind of enzymatic reaction, such as RT-PCR, TaqMan, Northern Blot or microarray analysis.

Kit components: NucleoSpin RNA Binding Plates, MN Wash Plates, Square-well Blocks, Round-well Blocks Low, Elution Plates U-bottom, Self-adhering Foil, RNase-free rDNase, RNase-free water and all buffers (Lysis RA1; Wash RA2, RA3 and RA4; Reaction for rDNase)

  • Sample material: <107 cultured cells (centrifugation), <30 mg tissue (centrifugation), saliva (collected with Oragene™)
  • Fragment size: >200 nt
  • Typical yield: 20 μg (2 x 106 HeLa cells), 20 μg (20 mg mouse liver)
  • A260/A280: 1.90–2.10
  • Typical RIN (RNA integrity number): >9 (cells), ≥7 (tissue)
  • RNA ratio: 28S/18S approx. 2.1
  • Typical concentration: 50–200ng/μL
  • Elution volume: 50–130 μL
  • Preparation time: 70 min./plate
  • Binding capacity: 100 μg

Specifications

Concentration 50–200ng/μL
Format 96-well Plate
Isolation Technology Silica Membrane
Elution Volume Elution volume: 50–130μL
Sample Type <1 x 107 Cultured Cells, <30 mg Tissue, Saliva (Collected with Oragene™)
For Use With (Application) Manual or automated isolation of total RNA from cultured cells and tissue
Includes NucleoSpin RNA Binding Plates (24), 1.5mL Collection Tubes (96), MN Wash Plates with 6 paper sheets (24), Square-well Blocks (36), Round-well Blocks Low with PE Foil (24), Elution Plates U-bottom with PE Foil (24), RNase-free rDNase (48 vials), Lysis Buffer RA1 (12 x 125mL), Wash Buffer RA2 (6 x 360mL), Wash Buffer RA3 Concentrate (12 x 100mL), Wash Buffer RA4 Concentrate (12 x 65mL), Reaction Buffer for rDNase (6 x 60mL), RNase-free Water (12 x 125mL)
No. of Reactions 24 x 96
Product Type NucleoSpin 96 RNA
Purity A260/A280: 1.90–2.10
Quantity 1 Kit
Sample Size <2 x 106 cultured cells, <20 mg human/animal tissue
Test Time 70 min/plate
Yield 20μg (2 × 106 HeLa cells)
20μg (20mg mouse liver)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoSpin 96 RNA
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash with water thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate/ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.